Assay Details
Gene Symbol: DENND2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098093.1 | NP_001091562.1 | 11 - 12 | 2519 | 58 | |
GenBank mRNA | BC140494.1 | - | 11 - 12 | 2519 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
