Assay Details
Gene Symbol: DTWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098120.2 | NP_001091589.1 | 1 - 2 | 96 | 126 | |
GenBank mRNA | BC140634.1 | - | 1 - 2 | 97 | 126 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC140634.1, NM_001098120.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
