Assay Details
Gene Symbol: DTWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098120.2 | NP_001091589.1 | 3 - 4 | 552 | 118 | |
GenBank mRNA | BC140634.1 | - | 3 - 4 | 553 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
