Assay Details
Gene Symbol: SHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098140.1 | NP_001091609.1 | 2 - 3 | 913 | 118 | |
GenBank mRNA | BC140673.1 | - | 2 - 3 | 913 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
