Assay Details
Gene Symbol: MPV17L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098145.2 | NP_001091614.1 | 2 - 3 | 385 | 93 | |
GenBank mRNA | BC140467.1 | - | 2 - 3 | 386 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
