Assay Details
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099036.1 | NP_001092506.1 | 3 - 4 | 1038 | 135 | |
GenBank mRNA | BC146083.1 | - | 3 - 4 | 1038 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
