Assay Details
Gene Symbol: PRSS54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099044.2 | NP_001092514.1 | 1 - 2 | 164 | 70 | |
GenBank mRNA | BC142059.1 | - | 1 - 2 | 165 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC142059.1, NM_001099044.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
