Assay Details
Gene Symbol: PRSS54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099044.2 | NP_001092514.1 | 5 - 6 | 924 | 77 | |
GenBank mRNA | BC142059.1 | - | 5 - 6 | 925 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
