Assay Details
Gene Symbol: DET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099117.1 | NP_001092587.1 | 2 - 3 | 1209 | 127 | |
GenBank mRNA | BC142507.1 | - | 2 - 3 | 1209 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
