Assay Details
Gene Symbol: ISL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099130.1 | NP_001092600.1 | 4 - 5 | 981 | 70 | |
GenBank mRNA | BC146163.1 | - | 4 - 5 | 981 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
