Assay Details
Gene Symbol: METTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099135.2 | NP_001092605.1 | 2 - 3 | 540 | 133 | |
GenBank mRNA | BC142084.1 | - | 2 - 3 | 541 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
