Assay Details
Gene Symbol: METTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099135.2 | NP_001092605.1 | 4 - 5 | 795 | 81 | |
GenBank mRNA | BC142084.1 | - | 4 - 5 | 796 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
