Assay Details
Gene Symbol: C1QTNF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099138.1 | NP_001092608.1 | 1 - 2 | 168 | 142 | |
GenBank mRNA | BC142162.1 | - | 1 - 2 | 168 | 142 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC142162.1, NM_001099138.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
