Assay Details
Gene Symbol: BET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099157.1 | NP_001092627.1 | 3 - 4 | 308 | 135 | |
GenBank mRNA | BC146171.1 | - | 3 - 4 | 308 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
