Assay Details
Gene Symbol: IGFBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099161.2 | NP_001092631.1 | 3 - 4 | 713 | 87 | |
GenBank mRNA | BC142451.1 | - | 3 - 4 | 714 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
