Assay Details
Gene Symbol: HOXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099174.1 | NP_001092644.1 | 1 - 2 | 508 | 142 | |
GenBank mRNA | BC142499.1 | - | 1 - 2 | 508 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
