Assay Details
Gene Symbol: RAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099179.2 | NP_001092649.1 | 3 - 4 | 304 | 59 | |
GenBank mRNA | BC142381.1 | - | 3 - 4 | 305 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
