Assay Details
Gene Symbol: C1QTNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101138.1 | NP_001094608.1 | 4 - 5 | 563 | 83 | |
GenBank mRNA | BC151389.1 | - | 4 - 5 | 563 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
