Assay Details
Gene Symbol: RPS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101152.2 | NP_001094622.1 | 1 - 2 | 32 | 67 | |
GenBank mRNA | BC147956.1 | - | 1 - 2 | 33 | 67 | |
BC148016.1 | - | 1 - 2 | 67 | |||
BC149631.1 | - | 1 - 2 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC147956.1, BC148016.1, NM_001101152.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
