Assay Details
Gene Symbol: RPS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101152.2 | NP_001094622.1 | 2 - 3 | 149 | 55 | |
GenBank mRNA | BC147956.1 | - | 2 - 3 | 150 | 55 | |
BC148016.1 | - | 2 - 3 | 151 | 55 | ||
BC149631.1 | - | 2 - 3 | 151 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
