Assay Details
Gene Symbol: EGLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101164.2 | NP_001094634.1 | 3 - 4 | 885 | 74 | |
GenBank mRNA | BC151442.1 | - | 3 - 4 | 886 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
