Assay Details
Gene Symbol: CNOT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101167.2 | NP_001094637.1 | 6 - 7 | 753 | 133 | |
GenBank mRNA | BC151334.1 | - | 6 - 7 | 754 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
