Assay Details
Gene Symbol: S1PR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101186.1 | NP_001094656.1 | 1 - 2 | 55 | 90 | |
GenBank mRNA | BC150076.1 | - | 1 - 2 | 55 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC150076.1, NM_001101186.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
