Assay Details
Gene Symbol: MANF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101211.2 | NP_001094681.1 | 3 - 4 | 396 | 106 | |
GenBank mRNA | BC149246.1 | - | 3 - 4 | 397 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
