Assay Details
Gene Symbol: EFCAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101247.1 | NP_001094717.1 | 4 - 5 | 301 | 115 | |
GenBank mRNA | BC149943.1 | - | 4 - 5 | 301 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
