Assay Details
Gene Symbol: VPS51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102093.2 | NP_001095563.1 | 5 - 6 | 1480 | 113 | |
GenBank mRNA | BC149640.1 | - | 5 - 6 | 1481 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
