Assay Details
Gene Symbol: BOLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102115.1 | NP_001095585.1 | 9 - 10 | 913 | 73 | |
NM_001168704.1 | NP_001162175.1 | 9 - 10 | 877 | 73 | ||
GenBank mRNA | BC149292.1 | - | 9 - 10 | 913 | 73 | |
EU050657.1 | - | 9 - 10 | 825 | 73 | ||
FJ623266.1 | - | 9 - 10 | 828 | 73 | ||
GU187434.1 | - | 9 - 10 | 877 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
