Assay Details
Gene Symbol: SCML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102184.2 | NP_001095654.1 | 1 - 2 | 192 | 67 | |
GenBank mRNA | BC149641.1 | - | 1 - 2 | 195 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC149641.1, NM_001102184.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
