Assay Details
Gene Symbol: FMR1NB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102262.2 | NP_001095732.1 | 2 - 3 | 509 | 88 | |
GenBank mRNA | BC149653.1 | - | 2 - 3 | 510 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
