Assay Details
Gene Symbol: RGCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102276.2 | NP_001095746.1 | 3 - 4 | 520 | 97 | |
GenBank mRNA | BC148118.1 | - | 3 - 4 | 521 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
