Assay Details
Gene Symbol: RGCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102276.2 | NP_001095746.1 | 4 - 5 | 580 | 108 | |
GenBank mRNA | BC148118.1 | - | 4 - 5 | 581 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
