Assay Details
Gene Symbol: PTP4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104998.1 | NP_001098468.1 | 4 - 5 | 674 | 73 | |
GenBank mRNA | BC147865.1 | - | 4 - 5 | 674 | 73 | |
BC151453.1 | - | 4 - 5 | 674 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
