Assay Details
Gene Symbol: ZNF346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105405.1 | NP_001098875.1 | 5 - 6 | 775 | 69 | |
GenBank mRNA | BC134498.1 | - | 5 - 6 | 775 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
