Assay Details
Gene Symbol: KIAA2012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105438.2 | NP_001098908.2 | 6 - 7 | 1271 | 68 | |
GenBank mRNA | BC149723.1 | - | 6 - 7 | 1272 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC149723.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
