Assay Details
Gene Symbol: KIAA2012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105438.2 | NP_001098908.2 | 8 - 9 | 1583 | 74 | |
GenBank mRNA | BC149723.1 | - | 8 - 9 | 1585 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
