Assay Details
Gene Symbol: ABHD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105441.1 | NP_001098911.1 | 2 - 3 | 1006 | 70 | |
GenBank mRNA | BC149237.1 | - | 2 - 3 | 1006 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
