Assay Details
Gene Symbol: ZNRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105443.2 | NP_001098913.1 | 1 - 2 | 788 | 92 | |
GenBank mRNA | BC142537.1 | - | 1 - 2 | 789 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
