Assay Details
Gene Symbol: MAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105633.1 | NP_001099103.1 | 4 - 5 | 444 | 65 | |
GenBank mRNA | BC151520.1 | - | 4 - 5 | 444 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
