Assay Details
Gene Symbol: GNAQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110002.1 | NP_001103472.1 | 3 - 4 | 700 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
