Assay Details
Gene Symbol: VCPKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110083.2 | NP_001103553.1 | 5 - 6 | 691 | 151 | |
GenBank mRNA | BC149565.1 | - | 5 - 6 | 692 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
