Assay Details
Gene Symbol: SYPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110088.1 | NP_001103558.1 | 1 - 2 | 64 | 61 | |
GenBank mRNA | BC112703.1 | - | 1 - 2 | 64 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
