Assay Details
Gene Symbol: DBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113321.1 | NP_001106792.1 | 3 - 4 | 312 | 137 | |
GenBank mRNA | BC114181.1 | - | 3 - 4 | 287 | 137 | |
M15886.1 | - | 3 - 4 | 312 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
