Assay Details
Gene Symbol: PNPLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099200.1 | NP_001092670.1 | 7 - 8 | 1921 | 115 | |
GenBank mRNA | BC142232.1 | - | 7 - 8 | 1921 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
