Assay Details
Gene Symbol: MRAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099393.1 | NP_001092863.1 | 2 - 3 | 200 | 121 | |
GenBank mRNA | BC146206.1 | - | 2 - 3 | 200 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
