Assay Details
Gene Symbol: DUSP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101294.2 | NP_001094764.2 | 1 - 2 | 696 | 57 | |
GenBank mRNA | BC148065.1 | - | 1 - 2 | 696 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
