Assay Details
Gene Symbol: ELOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102333.2 | NP_001095803.2 | 1 - 2 | 210 | 93 | |
GenBank mRNA | BC149490.1 | - | 1 - 2 | 82 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
