Assay Details
Gene Symbol: HSCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102340.1 | NP_001095810.1 | 5 - 6 | 656 | 81 | |
GenBank mRNA | BC148020.1 | - | 5 - 6 | 656 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
