Assay Details
Gene Symbol: NFATC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102536.1 | NP_001096006.1 | 5 - 6 | 2442 | 61 | |
GenBank mRNA | BC148981.1 | - | 5 - 6 | 2442 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
