Assay Details
Gene Symbol: CDC42SE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102537.1 | NP_001096007.1 | 2 - 3 | 240 | 150 | |
GenBank mRNA | BC147986.1 | - | 2 - 3 | 240 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC147986.1, NM_001102537.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
