Assay Details
Gene Symbol: CDC42SE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102537.1 | NP_001096007.1 | 3 - 4 | 581 | 96 | |
GenBank mRNA | BC147986.1 | - | 3 - 4 | 581 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC147986.1, NM_001102537.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
